Anti-ADM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
23781-1-AP-150UL
10082-536EA 402 USD
10082-536
Anti-ADM2 Rabbit Polyclonal Antibody
Antibodies
ADM2 also named as adrenomedullin 2 or IMDS is a 148 amino-acid secreted protein, which belongs to adrenomedullin family of calcitonin-related peptide hormones and is expressed in the esophagus, stomach, jejunum, ileum, ileocecum, ascending colon, transverse colon, descending colon and rectum. ADM2 activates the cAMP-dependent pathway and may play a role in regulating gastrointestinal and cardiovascular bioactivities. ADM2 play a role in the regulation of peripheral tissues by calcitonin receptor-like receptor.

IHC: Human Stomach Tissue, 1:20-1:200

Type: Primary
Antigen: ADM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR