Anti-CCDC27 Rabbit Polyclonal Antibody

Supplier: Proteintech
23769-1-AP-150UL
10084-178EA 402 USD
10084-178
Anti-CCDC27 Rabbit Polyclonal Antibody
Antibodies
CCDC27 also named as coiled coil domain containing 27 is a 656 amino acids protein, which contains a coiled-coil domain. CCDC27 expression is predominantly in male testis and lowly in other tissues. CCDC27 is a candidate gene for low bone mineral density. The function of CCDC27 is still non-known and need to be further studied.

Western Blot: Human Testis Tissue, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: CCDC27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR