Anti-UCP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23673-1-AP-150UL
10096-716EA 524.34 USD
10096-716
Anti-UCP1 Rabbit Polyclonal Antibody
Antibodies
UCP-1 (Mitochondrial uncoupling protein 1), is a mitochondrial transporter protein that creates proton leaks across the inner mitochondrial membrane, thus uncoupling oxidative phosphorylation from ATP synthesis. It has been identified a key molecule for metabolic thermogenesis to avoid an excess of fat accumulation. UCP-1 expression is usually restricted to brown adipose when induced by cold exposure and thyroid hormone.

Western Blot: Mouse brown adipose Tissue, 1:200-1:2000; IHC:rat Brain Tissue, 1:20-1:200

Type: Primary
Antigen: UCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR