Anti-RPS25 Rabbit Polyclonal Antibody

Supplier: Proteintech
23599-1-AP-150UL
10093-954EA 524.48 USD
10093-954
Anti-RPS25 Rabbit Polyclonal Antibody
Antibodies
Ribosomal protein S25 (RPS25) is a novel MDM2 interacting protein. It may be involved in viral replication of Dicistroviridae and hepatitis C viruses. RPS25 is overexpressed in human leukemia cells exhibiting adriamycin resistance. It may have a role in p53-mediated apoptosis and cell-cycle arrest. The expected MW of RPS25 is about 15-17kDa. Catalog# 23599-1-AP is a rabbit polyclonal antibody raised against full-length RPS25, and is recommended for ELISA, Western blotting and IP assays.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RPS25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR