Anti-GRB10 Rabbit Polyclonal Antibody

Supplier: Proteintech
23591-1-AP-150UL
10081-820EA 402 USD
10081-820
Anti-GRB10 Rabbit Polyclonal Antibody
Antibodies
Growth factor receptor-bound protein 10 (GRB10) is an adapter protein which modulates coupling of a number of cell surface receptor kinases with specific signaling pathways. GRB10 has three consensus domains including pleckstrin homology (PH) domain, SH2/SH3 domain and Ras-associating domain. By binding to kinases, GRB10 suppresses signals from activated receptors tyrosine kinases, including the insulin (INSR) and insulin-like growth factor (IGF1R) receptors. It may play a role in mediating insulin-stimulated ubiquitination of INSR, leading to proteasomal degradation.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: GRB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR