Anti-SCGB2A2 Rabbit Polyclonal Antibody

Supplier: Proteintech
23501-1-AP-150UL
10094-188EA 402 USD
10094-188
Anti-SCGB2A2 Rabbit Polyclonal Antibody
Antibodies
SCGB2A2, also known as mammaglobin A, is a member of the secretoglobin superfamily. It is a breast cancer-associated antigen almost exclusively over-expressed in primary and metastatic human breast cancers, making it a specific molecular marker and a potential therapeutic target for breast cancer. SCGB2A2 is a 93-amino acid protein with a calculated molecular weight of 10.5 kDa. This polyclonal antibody is raised against the C-terminal 71 amino acids of the protein encoded by a cDNA (MGC:71974; BC067220).

Western Blot: MCF-7 Cells, 1:200-1:1000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: SCGB2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR