Anti-LAMB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23498-1-AP-150UL
10089-480EA 402 USD
10089-480
Anti-LAMB1 Rabbit Polyclonal Antibody
Antibodies
Laminins are a family of basement membrane glycoproteins composed of three nonidentical chains, laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) . Laminins contribute to the structure of extracellular matrix and influence the behavior of associated cells, such as adhesion, differentiation, migration, phenotype stability, and resistance to anoikis . LAMB1 (laminin, beta 1) is expressed in most tissues that produce basement membranes, and is one of the 3 chains constituting laminin 1, the first laminin isolated from Engelbreth-Holm-Swarm (EHS) tumor. LAMB1 is also a subunit of laminin-2 (laminin-211 or merosin), laminin-6 (laminin-311 or K-laminin), laminin-8 (laminin-411), laminin-10 (laminin-511) and laminin-12 (laminin-213).

Western Blot:NIH/3T3 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: LAMB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR