Anti-ESF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23496-1-AP-150UL
10086-652EA 402 USD
10086-652
Anti-ESF1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:1000-1:10000; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: ESF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR