Anti-KHDC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23443-1-AP-150UL
10089-174EA 402 USD
10089-174
Anti-KHDC1 Rabbit Polyclonal Antibody
Antibodies
KHDC1, also named as C6orf148, belongs to the KHDC1 family. KHDC1 has two isoforms with calculated MW 27 kDa and 18 kDa. It’s a membrane protein. We got 35-40 kDa in our WB detection.

IHC: Human Liver Tissue, 1:20-1:200; Western Blot:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: KHDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR