Anti-CBX6 Rabbit Polyclonal Antibody

Supplier: Proteintech
23426-1-AP-150UL
10084-148EA 402 USD
10084-148
Anti-CBX6 Rabbit Polyclonal Antibody
Antibodies
The Chromobox domain (Cbx) gene family, consisting of Polycomb and Heterochromatin Protein 1 genes, involves in transcriptional repression, cell cycle regulation and chromatin remodeling . Cbx6, other named neuronal pentraxin receptor (NPR), first described as a putative integral membrane pentraxin that interacts with neuronal pentraxin 1 and 2, has also been classified as a Pc protein . NPR was determined to be a member of a distinct pentraxin protein subfamily, and found to form heteropentamer swithNP1 andNP2, followed by release from cell membranes .

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200

Type: Primary
Antigen: CBX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR