Anti-ADH7 Rabbit Polyclonal Antibody

Supplier: Proteintech
23425-1-AP-150UL
10082-528EA 402 USD
10082-528
Anti-ADH7 Rabbit Polyclonal Antibody
Antibodies
ADH7(Alcohol dehydrogenase class 4 mu/sigma chain) is also named as gastric alcohol dehydrogenase, retinol dehydrogenase and belongs to the zinc-containing alcohol dehydrogenase family. The ubiquitous expression pattern of ADH7 during embryogenesis has led to the notion that the enzymatic conversion of retinol to retinal occurs more or less uniformly and plays no role in the spatial or temporal regulation of RA synthesis during embryonic development. It has 2 isoforms produced by alternative splicing and can exsit as a homodimer.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: ADH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR