Anti-DOCK180 Rabbit Polyclonal Antibody

Supplier: Proteintech
23421-1-AP-150UL
10086-060EA 402 USD
10086-060
Anti-DOCK180 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: DOCK180
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR