Anti-BGLAP Rabbit Polyclonal Antibody

Supplier: Proteintech
23418-1-AP-150UL
10081-694EA 402 USD
10081-694
Anti-BGLAP Rabbit Polyclonal Antibody
Antibodies
IHC: Human osteosarcoma Tissue, 1:20-1:200

Type: Primary
Antigen: BGLAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR