Anti-FKBPL Rabbit Polyclonal Antibody

Supplier: Proteintech
23369-1-AP-150UL
10087-048EA 402 USD
10087-048
Anti-FKBPL Rabbit Polyclonal Antibody
Antibodies
FKBPL, also named as DIR1, NG7 and WISp39, has similarity to the immunophilin protein family, which plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBPL levels may be a prognostic indicator and determinant of response to endocrine therapy .It can be detected the band between 38 Kda and 48 Kda by western blot.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: FKBPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR