Anti-TMTC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23349-1-AP-150UL
10096-110EA 402 USD
10096-110
Anti-TMTC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse bladder Tissue, 1:200-1:2000

Type: Primary
Antigen: TMTC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR