Anti-EGF Rabbit Polyclonal Antibody

Supplier: Proteintech
23333-1-AP-150UL
10089-214EA 402 USD
10089-214
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-13 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR