Anti-IDH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23309-1-AP-150UL
10081-872EA 402 USD
10081-872
Anti-IDH1 Rabbit Polyclonal Antibody
Antibodies
IDH1, also named as PICD and IDP, belongs to the isocitrate and isopropylmalate dehydrogenases family. It is a common feature of a major subset of primary human brain cancers. Its subunit structure is homodimer.IDH1 mutation is always heterozygotic and IDH1 functions as a dimer, theoretically there will be 25% each wild type and mutant homo-dimers and 50% hetero-dimers present in the tumor cells. This antibody is specific to IDH1.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: IDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR