Anti-TAZ Rabbit Polyclonal Antibody

Supplier: Proteintech
23306-1-AP-150UL
10095-620EA 537.7 USD
10095-620
Anti-TAZ Rabbit Polyclonal Antibody
Antibodies
Western Blot: MDA-MB-453s Cells, 1:200-1:1000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TAZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR