Anti-KCNN4 Rabbit Polyclonal Antibody

Supplier: Proteintech
23271-1-AP-150UL
10088-198EA 402 USD
10088-198
Anti-KCNN4 Rabbit Polyclonal Antibody
Antibodies
The intermediate conductance calcium-activated potassium channel KCNN4, also known as KCa3.1, IK1, or SK4, is a member of the potassium channel KCNN family. KCNN4 is expressed in T cells, mast cells, macrophages, erythrocytes, vascular smooth muscle cells, airway smooth muscle cells, intestinal and airway epithelial cells . Functionally, KCNN4 plays a key role in calcium-dependent cell functions, such as proliferation, activation, and migration, in a broad range of cell types .

Western Blot: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: KCNN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR