Anti-ZNF311 Rabbit Polyclonal Antibody

Supplier: Proteintech
23243-1-AP-150UL
10097-334EA 402 USD
10097-334
Anti-ZNF311 Rabbit Polyclonal Antibody
Antibodies
Zinc finger protein 311 (ZNF311), also named Zinc finger protein zfp-31 (ZFP31), is involved in regulation of transcription. It belongs to the krueppel C2H2-type zinfinger protein family, and contains 14 C2H2-type zinc fingers and 1 KRAB domain.This antibody specifically reacts with ZNF311 protein.

Western Blot: HL-60 Cells, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: ZNF311
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR