Anti-ZGPAT Rabbit Polyclonal Antibody

Supplier: Proteintech
23203-1-AP-150UL
10097-282EA 402 USD
10097-282
Anti-ZGPAT Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:500-1:5000

Type: Primary
Antigen: ZGPAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR