Anti-MMACHC Rabbit Polyclonal Antibody

Supplier: Proteintech
23191-1-AP-150UL
10089-664EA 402 USD
10089-664
Anti-MMACHC Rabbit Polyclonal Antibody
Antibodies
IHC: Human Heart Tissue, 1:20-1:200; Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: MMACHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR