Anti-TMEM127 Rabbit Polyclonal Antibody

Supplier: Proteintech
23142-1-AP-150UL
10096-080EA 402 USD
10096-080
Anti-TMEM127 Rabbit Polyclonal Antibody
Antibodies
IP: HeLa Cells, 1:200-1:2000; IF: HeLa Cells, 1:10-1:100; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: TMEM127
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR