Anti-SALL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
23114-1-AP-150UL
10081-474EA 537.7 USD
10081-474
Anti-SALL2 Rabbit Polyclonal Antibody
Antibodies
Sall2 (SALL2, sal-like 2, HSAL2, p150(Sal2)) are mammalian homologs of the Drosophila region-specific home-otic gene spalt (sal), which encodes a zinc finger- containing transcription regulator. Mammalian Sall1 may mediate higher order chromatin structure and may be a component of a distinct heterochromatin-dependent silencing process. Sall2 is a p53-independent regulator of p21 and BAX, and can function in some cell types as a regulator of cell growth and survival. Human Sall2 is present in brain, heart, kidney and pancreas. This antibody is specific to the SALL2 protein.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: SALL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR