Anti-XRN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23108-1-AP-150UL
10097-194EA 402 USD
10097-194
Anti-XRN1 Rabbit Polyclonal Antibody
Antibodies
Exoribonuclease I (XRN1), also known as Sep1 or Rar5, is a 1,694-amino acid protein that functions as the major cytoplasmic 5 prime to 3 prime exoribonuclease and plays an important role in mRNA turnover. XRN1 may also function in the microtubular cytoskeleton as well as in DNA recombination and replication. XRN1 induces the expression of stress granules (SGs), cytoplasmic aggregates of stalled translational preinitiation complexes that accumulate during stress, and GW bodies/processing bodies (PBs), distinct cytoplasmic sites of mRNA degradation. There are several isoforms of XRN1 that are produced as a result of alternative splicing events.Loss of XRN1 markedly affects cell growth rates.This antibody is specific to the XRN1 protein.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: XRN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR