Anti-CD99 Rabbit Polyclonal Antibody

Supplier: Proteintech
23079-1-AP-150UL
10084-406EA 402 USD
10084-406
Anti-CD99 Rabbit Polyclonal Antibody
Antibodies
CD99, also known as MIC2, is a heavily O-glycosylated transmembrane protein involved in T-cell adhesion processes and in spontaneous rosette formation with erythrocytes. CD99 is broadly distributed on many cell types, with particularly strong expression on human cortical thymocytes, Ewing’s sarcoma cells and peripheral primitive neuroectodermal tumors . In normal cells, CD99 has been functionally implicated in cell adhesion, migration, apoptosis, differentiation, activation, and proliferation of lymphocytes and monocyte extravasation and transport of several transmembrane proteins . CD99 displays two surface isoforms generated by alternative splicing: a long 32 kDa and a short 28 kDa form .

Western Blot:U-937 Cells, 1:500-1:5000; IHC: Human pancreas Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: CD99
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR