Anti-SORCS1 Rabbit Polyclonal Antibody

Supplier: Proteintech
23002-1-AP-150UL
10081-524EA 402 USD
10081-524
Anti-SORCS1 Rabbit Polyclonal Antibody
Antibodies
SORCS1, also named as SorCS, belongs to the family of VPS10 domain-containing receptors. SorCS1 immunoreactivity is widespread in a population of neurons throughout the brain. Two different types of cellular localization were observed. Most SorCS1 immunoreactive neurons exhibit a punctate cytoplasmic staining which extends into the dendrites, whilst occassionally SorCS1 neuronal immunoreactivity is associated with the plasma membrane.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: SORCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR