Anti-CDKAL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22988-1-AP-150UL
10084-536EA 402 USD
10084-536
Anti-CDKAL1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:1000-1:10000

Type: Primary
Antigen: CDKAL1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR