Anti-CASP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22915-1-AP-150UL
10082-336EA 537.7 USD
10082-336
Anti-CASP1 Rabbit Polyclonal Antibody
Antibodies
CASP1(caspase-1) is also named as IL1BC, IL1BCE and belongs to the peptidase C14A family. It is a cysteine protease that regulates inflammatory processes through its capacity to process and activate the interleukin-1-beta (IL1B), IL18, and IL33 precursor proteins. The active caspase-1 can increase cellular membrane permeability and intracellular calcium levels, which facilitates lysosome exocytosis and release of host antimicrobial factors and microbial products. It has 5 isoforms produced by alternative splicing.

Western Blot:Jurkat Cells, 1:500-1:5000; IF:C6 Cells, 1:10-1:100

Type: Primary
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR