Anti-MYBPC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22900-1-AP-150UL
10090-674EA 402 USD
10090-674
Anti-MYBPC1 Rabbit Polyclonal Antibody
Antibodies
MYBPC1, also named as MYBPCS, belongs to the immunoglobulin superfamily and MyBP family. It is a thick filament-associated protein located in the crossbridge region of vertebrate striated muscle a bands. In vitro it binds MHC, F-actin and native thin filaments, and modifies the activity of actin-activated myosin ATPase. It may modulate muscle contraction or may play a more structural role. This antibody is specific to MYBPC1.

  • Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: MYBPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR