Anti-HSF4 Rabbit Polyclonal Antibody

Supplier: Proteintech
22883-1-AP-150UL
10081-212EA 402 USD
10081-212
Anti-HSF4 Rabbit Polyclonal Antibody
Antibodies
HSF4 belongs to the HSF family. HSF4 is a DNA-binding protein that specifically binds heat shock promoter elements (HSE). The isoform HSF4A represses transcription while the isoform HSF4B activates transcription. HSF4 is a homotrimer which exhibits constitutive DNA binding and forms trimers even in the absence of stress. It is expressed in heart, skeletal muscle, eye and brain, and at much lower levels in some other tissues. Mutation of HSF4 will cause lamellar cataract which is known as Marner type cataract (CAM). Lamellar cataract is an autosomal dominant common type of infantile cataract. Finger malformation is observed in some kindreds. Inheritance of these forms of cataract is autosomal dominant.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: HSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR