Anti-MEX3C Rabbit Polyclonal Antibody

Supplier: Proteintech
22882-1-AP-150UL
10090-170EA 402 USD
10090-170
Anti-MEX3C Rabbit Polyclonal Antibody
Antibodies
MEX3C(RNA-binding E3 ubiquitin-protein ligase MEX3C) is also named as RKHD2, RNF194 and may be involved in post-transcriptional regulatory mechanisms. MEX3C is necessary for normal postnatal growth and enhances the expression of local bone Igf1 expression. This protein is expressed in the cytoplasm and shuttles between the cytoplasm and the nucleus through the CRM1 export pathway.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: MEX3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR