Anti-CSAD Rabbit Polyclonal Antibody

Supplier: Proteintech
22870-1-AP-150UL
10085-232EA 402 USD
10085-232
Anti-CSAD Rabbit Polyclonal Antibody
Antibodies
IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: CSAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR