Anti-IL22RA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22861-1-AP-150UL
10088-818EA 402 USD
10088-818
Anti-IL22RA2 Rabbit Polyclonal Antibody
Antibodies
Interleukin-22 receptor subunit alpha-2 (IL22RA2) is a soluble member of the type II cytokine receptor family. Through binding to IL22, IL22RA2 prevents the interaction of IL22 with the functional IL22-R complex and blocks the activity of IL22 (in vitro). IL22RA2 may play an important role as an IL22 antagonist in the regulation of inflammatory responses. Alternative splicing results in multiple transcript variants encoding distinct isoforms.

Western Blot: MDA-MB-453s Cells, 1:200-1:2000

Type: Primary
Antigen: IL22RA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR