Anti-OFD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22851-1-AP-150UL
10091-554EA 402 USD
10091-554
Anti-OFD1 Rabbit Polyclonal Antibody
Antibodies
OFD1,also named as CXorf5 and 71-7A, belongs to the OFD1 family. It has been implicated in several developmental syndromes, including a male-lethal X-linked dominant condition,Oral-Facial-Digital type 1 (OFD1) syndrome, X-linked recessiveSimpson-Golabi-Behmel syndrome type 2 (SGBS2) and Joubert syndrome and related disorders (JSRDs). It is a component of the centrioles controlling mother and daughter centrioles length. It is recruits to the centriole IFT88 and centriole distal appendage-specific proteins including CEP164. Involved in the biogenesis of the cilium, a centriole-associated function. OFD1 plays an important role in development by regulating Wnt signaling and the specification of the left-right axis. This antibody recognize all the isoforms of OFD1. The 43 kDa is isoform 2. The ~80 kDa band is unknown.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: OFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR