Anti-NO66/C14orf169 Rabbit Polyclonal Antibody

Supplier: Proteintech
22842-1-AP-150UL
10091-178EA 402 USD
10091-178
Anti-NO66/C14orf169 Rabbit Polyclonal Antibody
Antibodies
NO66, also known as C14orf169 and MAPJD, is a JmjC domain-containing oxygenase that can act as both a histone lysine demethylase and a ribosomal histidine hydroxylase. NO66 is highly conserved during evolution and shows a dual localization pattern, present in both the nucleoplasm and nucleolus . It has been reported that NO66 participates in regulation of Osx transcriptional activity and plays an important role in osteoblast differentiation through interaction with Osx . Through interaction with MYC protein, NO66 plays a role in pulmonary carcinogenesis .

Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: NO66/C14orf169
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR