Anti-CCS Rabbit Polyclonal Antibody

Supplier: Proteintech
22802-1-AP-150UL
10084-240EA 402 USD
10084-240
Anti-CCS Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:500-1:5000; IP: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: CCS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR