Anti-MIXL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22772-1-AP-150UL
10090-246EA 537.7 USD
10090-246
Anti-MIXL1 Rabbit Polyclonal Antibody
Antibodies
Mix1 homeobox-like 1 (MIXL1) Transcription factor that play a central role in proper axial mesendoderm morphogenesis and endoderm formation. Required for efficient differentiation of cells from the primitive streak stage to blood, by acting early in the recruitment and/or expansion of mesodermal progenitors to the hemangioblastic and hematopoietic lineages. Also involved in the morphogenesis of the heart and the gut during embryogenesis. Acts as a negative regulator of brachyury expression. This antibody is specific to MIXL1.

Western Blot: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: MIXL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR