Anti-RWDD3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22737-1-AP-150UL
10093-734EA 402 USD
10093-734
Anti-RWDD3 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: RWDD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR