Anti-Collagen Type III Rabbit Polyclonal Antibody

Supplier: Proteintech
22734-1-AP-150UL
10082-030EA 402 USD
10082-030
Anti-Collagen Type III Rabbit Polyclonal Antibody
Antibodies
Type III collagen is a fibrillar forming collagen comprising three a1(III) chains and is expressed in early embryos and throughout embryogenesis. In the adult, type III collagen is a major component of the extracellular matrix in a variety of internal organs and skin. It occurs in most soft connective tissues along with type I collagen. COL3A1 gene encodes type III procollagen. Mutations in this gene are associated with type III and IV Ehlers-Danlos syndrome, and with aortic and arterial aneurysms.

IHC: Human Colon Tissue, 1:20-1:200; Western Blot: Mouse skin Tissue, 1:200-1:1000

Type: Primary
Antigen: Collagen Type III
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR