Anti-NUAK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22723-1-AP-150UL
10083-114EA 537.7 USD
10083-114
Anti-NUAK1 Rabbit Polyclonal Antibody
Antibodies
NUAK1 is a member of the human adenosine monophosphate (AMP)-activated protein kinase family that has been identified as a key tumor cell survival factor. It is also named as ARK5, KIAA0537, OMPHK1 and belongs to the protein kinase superfamily. The physiological role of NUAK1 is potentially to suppress glucose uptake through negative regulation of insulin signaling in oxidative muscle. This protein has 2 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: NUAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR