Anti-AARS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22696-1-AP-150UL
10082-282EA 402 USD
10082-282
Anti-AARS2 Rabbit Polyclonal Antibody
Antibodies
AARS2(Alanine--tRNA ligase, mitochondrial) is also named as KIAA1270, AARSL, bA444E17.1, AlaRS, AARSL, COXPD8, MTALARS, MT-ALARS and belongs to the class-II aminoacyl-tRNA synthetase family. It catalyzes the attachment of alanine to tRNA(Ala) in a two-step reaction: alanine is first activated by ATP to form Ala-AMP and then transferred to the acceptor end of tRNA(Ala). It also edits incorrectly charged tRNA(Ala) via its editing domain. The full length 107 kDa protein has a transit peptide with 23 amino acids.

Western Blot:K-562 Cells, 1:1000-1:10000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: AARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR