Anti-PPP4R2 Rabbit Polyclonal Antibody

Supplier: Proteintech
22676-1-AP-150UL
10092-732EA 402 USD
10092-732
Anti-PPP4R2 Rabbit Polyclonal Antibody
Antibodies
PPP4R2 is a regulatory subunit of protein phosphatase-4 (PPP4), a protein serine/threonine phosphatase implicated in microtubule organization at centrosomes and DNA double-strand break repair. The deduced 453-amino acid protein contains several potential phosphorylation sites and has a calculated molecular mass of 50.4 kD. Immunofluorescent analysis of HeLa and A431 cells revealed prominent localization of PPP4R2 at centrosomes, with weaker staining of cytoplasm and nucleus.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PPP4R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR