Anti-SART1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22675-1-AP-150UL
10094-146EA 402 USD
10094-146
Anti-SART1 Rabbit Polyclonal Antibody
Antibodies
Squamous cell carcinoma antigen recognized by T cells (SART1), also known as U4/U6.U5 tri-snRNP-associated 110 kDa protein, is a ubiquitously expressed protein that is involved in mRNA splicing and the regulation of cell proliferation. It has been identified as regulator of c-FLIP and drug-induced activation of caspase 8. The SART1 gene encodes two proteins, the SART1(800) protein is located in the nucleus of the majority of proliferating cells, while the SART1(259) protein is located in the cytosol of epithelial cancers. Genetic variation in the SART1 gene may be associated with breast cancer development. This antibody recognizes the endogenous 800-amino acid SART1, which migrates with an apparent molecular mass of 110 kDa in SDS-PAGE.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: SART1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR