Anti-FSHR Rabbit Polyclonal Antibody

Supplier: Proteintech
22665-1-AP-150UL
10087-164EA 402 USD
10087-164
Anti-FSHR Rabbit Polyclonal Antibody
Antibodies
Western Blot:DU 145 Cells, 1:500-1:5000

Type: Primary
Antigen: FSHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR