Anti-EYA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
22658-1-AP-150UL
10086-736EA 402 USD
10086-736
Anti-EYA1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Kidney Tissue, 1:20-1:200; Western Blot: Mouse Kidney Tissue, 1:200-1:2000

Type: Primary
Antigen: EYA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR