Anti-DNASE2B Rabbit Polyclonal Antibody

Supplier: Proteintech
22652-1-AP-150UL
10086-036EA 402 USD
10086-036
Anti-DNASE2B Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:1000-1:10000; IF:A549 Cells, 1:10-1:100; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: DNASE2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR