Anti-C9orf72 Rabbit Polyclonal Antibody

Supplier: Proteintech
22637-1-AP-150UL
10083-902EA 524.48 USD
10083-902
Anti-C9orf72 Rabbit Polyclonal Antibody
Antibodies
C9ORF72 has a domain whith polymorphic hexanucleotide repeat (GGGGCC). The C9ORF72-hexanucleotide repeat expansions have been recently identified as genetic markers in amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). The C9ORF72 repeat expansions may indicate a worse prognosis in ALS. C9ORF72 has two isoforms with MW 54-60 kDa and 25-30 kDa.

Western Blot:SH-SY5Y Cells, 1:200-1:1000; IHC: Human Brain Tissue, 1:20-1:200; IF:SH-SY5Y Cells, 1:10-1:100; IP:SH-SY5Y Cells, 1:200-1:1000

Type: Primary
Antigen: C9orf72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR