Anti-ZZZ3 Rabbit Polyclonal Antibody

Supplier: Proteintech
22608-1-AP-150UL
10097-416EA 402 USD
10097-416
Anti-ZZZ3 Rabbit Polyclonal Antibody
Antibodies
Zinc finger, ZZ-type containing 3 (ZZZ3) is a component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.

Western Blot: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: ZZZ3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR