Anti-BCR Rabbit Polyclonal Antibody

Supplier: Proteintech
22585-1-AP-150UL
10083-576EA 402 USD
10083-576
Anti-BCR Rabbit Polyclonal Antibody
Antibodies
The Bcr gene, mapping on chromosome22, was orginally identified by its presence in the chimeric Bcr-Abl oncogene. The Bcr gene encodes for the break point cluster region protein(Bcr). Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for RAC1 and CDC42. A chromosomal aberration involving BCR has been found in patients with chronic myeloid leukemia.

Western Blot:Y79 Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: BCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR